accessibility menu, dialog, popup

UserName

CAS RN 94280-72-5

CH3NCH3Br

CAS RN 94280-72-5

IUPAC Name: 1-butyl-1-methylpiperidin-1-ium bromide
Synonyms: 1-butyl-1-methylpiperidin-1-ium bromide
Molecular Weight (g/mol): 236.2
Molecular Formula: C10H22BrN
InChi Key: WYABBCZMFVULEF-UHFFFAOYSA-M
SMILES: [Br-].CCCC[N+]1(C)CCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results