accessibility menu, dialog, popup

UserName

Explore All Our Special Offers, All in One Place

Save Today

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 94242-85-0

BOOH3CCH3H3CH3CCH3

CAS RN 94242-85-0

IUPAC Name: pentamethyl-1,3,2-dioxaborolane
Synonyms: pentamethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 142.01
Molecular Formula: C7H15BO2
InChi Key: FOQJHZPURACERJ-UHFFFAOYSA-N
SMILES: CB1OC(C)(C)C(C)(C)O1
Boiling Point 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.