missing translation for 'onlineSavingsMsg'
Learn More

2,4,4,5,5-Pentamethyl-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. P26025G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P26025G 5 g
P26021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P26025G Supplier TCI America Supplier No. P26025G
Only null left

Chemical Identifiers

CAS 94242-85-0
Molecular Formula C7H15BO2
Molecular Weight (g/mol) 142.005
MDL Number MFCD09027082
InChI Key FOQJHZPURACERJ-UHFFFAOYSA-N
PubChem CID 11759390
IUPAC Name 2,4,4,5,5-pentamethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C

Specifications

Color Colorless
Boiling Point 35°C
Quantity 5 g
UN Number 3272
Formula Weight 142.01
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,4,5,5-Pentamethyl-1,3,2-dioxaborolane
TSCA No
Recommended Storage Refrigerator