CAS RN 942-01-8
CAS RN 942-01-8
IUPAC Name:
2,3,4,9-tetrahydro-1H-carbazole
Synonyms:
2,3,4,9-tetrahydro-1H-carbazole
Molecular Weight (g/mol):
171.24
Molecular Formula:
C12H13N
InChi Key:
XKLNOVWDVMWTOB-UHFFFAOYSA-N
SMILES:
C1CCC2=C(C1)NC1=CC=CC=C21
1
–
1
of
1
results
1
1,2,3,4-Tetrahydrocarbazole 99.0+%, TCI America™
CAS: 942-01-8 Molecular Formula: C12H13N Molecular Weight (g/mol): 171.243 MDL Number: MFCD00004959 InChI Key: XKLNOVWDVMWTOB-UHFFFAOYSA-N Synonym: 1,2,3,4-tetrahydrocarbazole,2,3-tetramethyleneindole,2,3-tetramethylene-1h-indole,5,6,7,8-tetrahydrocarbazole,tetrahydrocarbazole,1h-carbazole, 2,3,4,9-tetrahydro,carbazole, 1,2,3,4-tetrahydro,1h-indole, 2,3-1,4-butanediyl,carbazole, 5,6,7,8-tetrahydro,unii-8zlk0tsx93 PubChem CID: 13664 IUPAC Name: 2,3,4,9-tetrahydro-1H-carbazole SMILES: C1CCC2=C(C1)C3=CC=CC=C3N2