accessibility menu, dialog, popup

UserName

ClNHNN

CAS RN 94-97-3

IUPAC Name: 5-chloro-2H-1,2,3-benzotriazole
Synonyms: 5-chloro-2H-1,2,3-benzotriazole
Molecular Weight (g/mol): 153.57
Molecular Formula: C6H4ClN3
InChi Key: PZBQVZFITSVHAW-UHFFFAOYSA-N
SMILES: ClC1=CC2=NNN=C2C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results