missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorobenzotriazole 98.0+%, TCI America™

Catalog No. C013725G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C013725G 25 g
C0137100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C013725G Supplier TCI America Supplier No. C013725G
Only null left

Chemical Identifiers

CAS 94-97-3
Molecular Formula C6H4ClN3
Molecular Weight (g/mol) 153.57
MDL Number MFCD00005700
InChI Key PZBQVZFITSVHAW-UHFFFAOYSA-N
Synonym 5-chlorobenzotriazole, 1h-benzotriazole, 5-chloro, 6-chlorobenzotriazole, 5-chloro-1h-benzotriazole, 1h-benzotriazole, 6-chloro, 5-chloro-1h-benzo d 1,2,3 triazole, 5-chloro-1h-1,2,3-benzotriazole, 6-chloro-1h-benzotriazole, unii-8c7o46g78k, 5-chlorobenztriazole
PubChem CID 66760
IUPAC Name 5-chloro-2H-1,2,3-benzotriazole
SMILES ClC1=CC2=NNN=C2C=C1

Specifications

Melting Point 158°C
Color White-Yellow
Quantity 25 g
Formula Weight 153.57
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chlorobenzotriazole
EINECSNumber (5)-0540
TSCA Yes