accessibility menu, dialog, popup

UserName

H3CHNOOCH3

CAS RN 94-32-6

IUPAC Name: ethyl 4-(butylamino)benzoate
Synonyms: ethyl 4-(butylamino)benzoate
Molecular Weight (g/mol): 221.3
Molecular Formula: C13H19NO2
InChi Key: GTXRSQYDLPYYNW-UHFFFAOYSA-N
SMILES: CCCCNC1=CC=C(C=C1)C(=O)OCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results