missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-(Butylamino)benzoate 98.0+%, TCI America™

Catalog No. B147225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B147225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B147225G Supplier TCI America Supplier No. B147225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 94-32-6
Molecular Formula C13H19NO2
Molecular Weight (g/mol) 221.30
MDL Number MFCD00017283
InChI Key GTXRSQYDLPYYNW-UHFFFAOYSA-N
Synonym ethyl 4-butylamino benzoate, ethyl p-butylaminobenzoate, benzoic acid, 4-butylamino-, ethyl ester, unii-d68ep3v31w, 4-butylamino benzoic acid ethyl ester, 4-n-butylamino benzoic acid ethyl ester, acmc-209rrf, ethyl4-butylamino benzoate, ethyl p-butylamino benzoate, ethyl 4-n-butylamino benzoate
PubChem CID 66746
IUPAC Name ethyl 4-(butylamino)benzoate
SMILES CCCCNC1=CC=C(C=C1)C(=O)OCC

Specifications

Melting Point 70°C
Color White
Boiling Point 220°C
Quantity 25 g
Formula Weight 221.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-(Butylamino)benzoate