accessibility menu, dialog, popup

UserName

CAS RN 939-97-9

CH3H3CCH3O

CAS RN 939-97-9

IUPAC Name: 4-tert-butylbenzaldehyde
Synonyms: 4-T-butyl benzaldehyde
Molecular Weight (g/mol): 162.23
Molecular Formula: C11H14O
InChi Key: OTXINXDGSUFPNU-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=O)C=C1
Boiling Point 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)