missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzaldehyde, 97%

Catalog No. AAA1574022
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1574022 100 g
AAA1574006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1574022 Supplier Thermo Scientific Chemicals Supplier No. A1574022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 939-97-9 | C11H14O | 162.23 g/mol

Used for preparing isoxazolyl penicillin derivatives. 4-tert-Butylbenzaldehyde is suitable for use in a kinetic study to evaluate the kinetic constants (KI) for inhibition of mushroom tyrosinase by 4-substituted benzaldehydes. It is also an important intermediate for the synthesis of medicines, dyes, flavor and fragrance compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used for preparing isoxazolyl penicillin derivatives. 4-tert-Butylbenzaldehyde is suitable for use in a kinetic study to evaluate the kinetic constants (KI) for inhibition of mushroom tyrosinase by 4-substituted benzaldehydes. It is also an important intermediate for the synthesis of medicines, dyes, flavor and fragrance compounds.

Solubility
Soluble in water.

Notes
Air and light sensitive. Store away from air, light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.

Chemical Identifiers

CAS 939-97-9
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00035742
InChI Key OTXINXDGSUFPNU-UHFFFAOYSA-N
Synonym 4-tert-butyl benzaldehyde, p-t-butylbenzaldehyde, p-tertbutyl benzaldehyde, benzaldehyde, 4-1,1-dimethylethyl, p-tert-butylbenzaldehyde, 4-t-butylbenzaldehyde, 4-tert-butyl-benzaldehyde, unii-qxf0qy8503, 4-t-butyl benzaldehyde, benzaldehyde, p-tert-butyl
PubChem CID 70324
IUPAC Name 4-tert-butylbenzaldehyde
SMILES CC(C)(C)C1=CC=C(C=O)C=C1

Specifications

Density 0.97
Boiling Point 244°C to 246°C
Flash Point 101°C (213°F)
Refractive Index 1.5285
Quantity 100 g
UN Number UN3082
Beilstein 1906461
Sensitivity Air and light sensitive
Solubility Information Soluble in water.
Formula Weight 162.23
Percent Purity 97%
Chemical Name or Material 4-tert-Butylbenzaldehyde
Show More Show Less
Hazard Category H317-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P271-P272-P280g-P302+P352-P304+P340-P312-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 213-367-9
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only