accessibility menu, dialog, popup

UserName

CAS RN 938-96-5

CH3(S)OOOH

CAS RN 938-96-5

IUPAC Name: (2S)-2-(4-hydroxyphenyl)propanoate
Synonyms: (2S)-2-(4-hydroxyphenyl)propanoate
Molecular Weight (g/mol): 165.17
Molecular Formula: C9H9O3
InChi Key: ZHMMPVANGNPCBW-LURJTMIESA-M
SMILES: C[C@H](C([O-])=O)C1=CC=C(O)C=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results