missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Hydroxyphenyl)propionic Acid 98.0+%, TCI America™

Catalog No. H103725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H103725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H103725G Supplier TCI America Supplier No. H103725G

Chemical Identifiers

CAS 938-96-5
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00128847
InChI Key ZHMMPVANGNPCBW-UHFFFAOYSA-N
Synonym 2-4-hydroxyphenyl propanoic acid, 2-4-hydroxyphenyl propionic acid, 4-hydroxyhydratropate, 4-hydroxyhydratropic acid, 4-hydroxyphenyl-2-propionic acid, 2-4'-hydroxyphenyl-propionic acid, 2-4-hydroxyphenyl propionate, acmc-209ro0, 4-hydroxyphenyl propanoic acid, methyl 4-hydroxyphenylacetic acid
PubChem CID 102526
ChEBI CHEBI:1868
IUPAC Name 2-(4-hydroxyphenyl)propanoic acid
SMILES CC(C1=CC=C(C=C1)O)C(=O)O

Specifications

Melting Point 131°C
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Hydroxyphenyl)propionic Acid