accessibility menu, dialog, popup

UserName

CAS RN 938-09-0

IUPAC Name: (2-chloroethanesulfonyl)benzene
Synonyms: (2-chloroethanesulfonyl)benzene
Molecular Weight (g/mol): 204.67
Molecular Formula: C8H9ClO2S
InChi Key: NUJGORANFDSMOL-UHFFFAOYSA-N
SMILES: ClCCS(=O)(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)