accessibility menu, dialog, popup

UserName

FFFHS

CAS RN 937-00-8

IUPAC Name: 3-(trifluoromethyl)benzene-1-thiol
Synonyms: 3-(trifluoromethyl)benzenethiol
Molecular Weight (g/mol): 178.17
Molecular Formula: C7H5F3S
InChi Key: SCURCOWZQJIUGR-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(S)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)