CAS RN 936563-96-1
IUPAC Name:
1-{3-[4-amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]piperidin-1-yl}prop-2-en-1-one
Synonyms:
ibrutinib
Molecular Weight (g/mol):
440.51
Molecular Formula:
C25H24N6O2
InChi Key:
XYFPWWZEPKGCCK-UHFFFAOYNA-N
SMILES:
NC1=C2C(=NC=N1)N(N=C2C1=CC=C(OC2=CC=CC=C2)C=C1)C1CCCN(C1)C(=O)C=C
1
–
2
of
2
results
1
Promotions Available
Thermo Scientific Chemicals Ibrutinib
Thermo Scientific Chemicals Ibrutinib
CAS: 936563-96-1 Molecular Formula: C25H24N6O2
2
Tocris Bioscience™ Ibrutinib
Potent and selective BTK inhibitor (IC50= 0.5 nM)