missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ibrutinib

Catalog No. AC466802500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466802500 250 mg
AC466800010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466802500 Supplier Thermo Scientific Chemicals Supplier No. 466802500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 936563-96-1 | C25H24N6O2, C25H24N6O2 | 440.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 936563-96-1
Molecular Formula C25H24N6O2

Specifications

Quantity 250 mg
Formula Weight 440.51
Chemical Name or Material Ibrutinib
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement May damage fertility or the unborn child.

May cause damage to organs through prolonged or repeated exposure.

Precautionary Statement GHS P Statement Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only