accessibility menu, dialog, popup

UserName

OOO

CAS RN 935-13-7

IUPAC Name: 3-(furan-2-yl)propanoate
Synonyms: 3-(furan-2-yl)propanoate
Molecular Weight (g/mol): 139.13
Molecular Formula: C7H7O3
InChi Key: XLTJXJJMUFDQEZ-UHFFFAOYSA-M
SMILES: [O-]C(=O)CCC1=CC=CO1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)