missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Furyl)propionic acid, 97%

Catalog No. AAH2727306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2727306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2727306 Supplier Thermo Scientific Chemicals Supplier No. H2727306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 935-13-7 | C7H8O3 | 140.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 935-13-7
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00005346
InChI Key XLTJXJJMUFDQEZ-UHFFFAOYSA-N
Synonym 3-furan-2-yl propanoic acid, 3-2-furyl propanoic acid, 2-furanpropanoic acid, 2-furanpropionic acid, 3-2-furyl propionic acid, 3-furan-2-yl-propionic acid, furan-2-propionic acid, 3-2-furyl propoinic acid, furfuryl-essigsaure, furfurylacetic acid
PubChem CID 70286
IUPAC Name 3-(furan-2-yl)propanoic acid
SMILES C1=COC(=C1)CCC(=O)O

Specifications

Melting Point 56°C to 60°C
Quantity 5 g
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material 3-(2-Furyl)propionic acid
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 213-298-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only