accessibility menu, dialog, popup

UserName

H3CONOH

CAS RN 93413-76-4

IUPAC Name: 2-(1-hydroxycyclohexyl)-2-(4-methoxyphenyl)acetonitrile
Synonyms: 2-(1-hydroxycyclohexyl)-2-(4-methoxyphenyl)acetonitrile
Molecular Weight (g/mol): 245.32
Molecular Formula: C15H19NO2
InChi Key: ASYJSBPNAIDUHX-UHFFFAOYNA-N
SMILES: COC1=CC=C(C=C1)C(C#N)C1(O)CCCCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results