missing translation for 'onlineSavingsMsg'
Learn More

(1-Hydroxycyclohexyl)(4-methoxyphenyl)acetonitrile 98.0+%, TCI America™

Catalog No. H13031G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H13031G Supplier TCI America Supplier No. H13031G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 93413-76-4
Molecular Formula C15H19NO2
Molecular Weight (g/mol) 245.322
MDL Number MFCD06658142
InChI Key ASYJSBPNAIDUHX-UHFFFAOYSA-N
Synonym 1-[Cyano(4-methoxyphenyl)methyl]cyclohexanol
PubChem CID 9899621
IUPAC Name 2-(1-hydroxycyclohexyl)-2-(4-methoxyphenyl)acetonitrile
SMILES COC1=CC=C(C=C1)C(C#N)C2(CCCCC2)O

Specifications

Melting Point 127°C
Color White
Quantity 1 g
Formula Weight 245.32
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (1-Hydroxycyclohexyl)(4-methoxyphenyl)acetonitrile
TSCA No
Recommended Storage Refrigerator