accessibility menu, dialog, popup

UserName

OHNS

CAS RN 934-34-9

IUPAC Name: 2,3-dihydro-1,3-benzothiazol-2-one
Synonyms: 2-benzothiazolone
Molecular Weight (g/mol): 151.18
Molecular Formula: C7H5NOS
InChi Key: YEDUAINPPJYDJZ-UHFFFAOYSA-N
SMILES: O=C1NC2=CC=CC=C2S1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)