missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzothiazole, 98%

Catalog No. AAA1664814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1664814 25 g
AAA1664806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1664814 Supplier Thermo Scientific Chemicals Supplier No. A1664814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 934-34-9 | C7H5NOS | 151.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 934-34-9
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.183
MDL Number MFCD00022868
InChI Key YEDUAINPPJYDJZ-UHFFFAOYSA-N
Synonym benzothiazolone, 2-hydroxybenzothiazole, 2 3h-benzothiazolone, 2-benzothiazolol, 2-benzothiazolone, 1,3-benzothiazol-2-ol, 2-benzothiazolinone, 3h-benzothiazol-2-one, benzothiazolol, benzo d thiazol-2 3h-one
PubChem CID 13625
ChEBI CHEBI:115196
IUPAC Name 3H-1,3-benzothiazol-2-one
SMILES C1=CC=C2C(=C1)NC(=O)S2

Specifications

Melting Point 136°C to 140°C
Quantity 25 g
Formula Weight 151.18
Percent Purity 98%
Physical Form Crystals or Powder or Crystalline Powder
Chemical Name or Material 2-Hydroxybenzothiazole
Hazard Category Warning
Hazard Statement GHS H Statement

H315-H319-H335

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement GHS P Statement

P261-P280-P305+P351+P338-P304+P340-P405-P501a

Avoid breathing dust/fume/gas/mist/vapors/spray.

Wear protective gloves/protective clothing/eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Store locked up.

Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 213-281-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only