accessibility menu, dialog, popup

UserName

ONNH2F

CAS RN 93246-53-8

IUPAC Name: 3-fluoro-4-(morpholin-4-yl)aniline
Synonyms: 3-fluoro-4-(morpholin-4-yl)aniline
Molecular Weight (g/mol): 196.23
Molecular Formula: C10H13FN2O
InChi Key: FQGIBHQUVCGEAC-UHFFFAOYSA-N
SMILES: NC1=CC(F)=C(C=C1)N1CCOCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results