missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-morpholinoaniline 98.0+%, TCI America™

Catalog No. F08495G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08495G 5 g
F084925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08495G Supplier TCI America Supplier No. F08495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 93246-53-8
Molecular Formula C10H13FN2O
Molecular Weight (g/mol) 196.23
MDL Number MFCD02571270
InChI Key FQGIBHQUVCGEAC-UHFFFAOYSA-N
Synonym 4-(2-Fluoro-4-aminophenyl)morpholine
PubChem CID 1485330
IUPAC Name 3-fluoro-4-(morpholin-4-yl)aniline
SMILES NC1=CC(F)=C(C=C1)N1CCOCC1

Specifications

Melting Point 123°C
Quantity 5 g
Formula Weight 196.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-4-morpholinoaniline