accessibility menu, dialog, popup

UserName

BrCl

CAS RN 932-77-4

IUPAC Name: 1-bromo-3-(chloromethyl)benzene
Synonyms: 1-bromo-3-(chloromethyl)benzene
Molecular Weight (g/mol): 205.48
Molecular Formula: C7H6BrCl
InChi Key: UDKGXKYEWBGQCG-UHFFFAOYSA-N
SMILES: ClCC1=CC(Br)=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)