accessibility menu, dialog, popup

UserName

H3CNHCl

CAS RN 932-32-1

IUPAC Name: 2-chloro-N-methylaniline
Synonyms: 2-chloro-N-methylaniline
Molecular Weight (g/mol): 141.6
Molecular Formula: C7H8ClN
InChi Key: WGNNILPYHCKCFF-UHFFFAOYSA-N
SMILES: CNC1=CC=CC=C1Cl
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)