accessibility menu, dialog, popup

UserName

CAS RN 93164-73-9

IUPAC Name: 5-octyl-2,2'-bithiophene
Synonyms: 5-octyl-2,2'-bithiophene
Molecular Weight (g/mol): 278.47
Molecular Formula: C16H22S2
InChi Key: ZYEIXHZLTHTEOI-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=CC=C(S1)C1=CC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results