missing translation for 'onlineSavingsMsg'
Learn More

5-n-Octyl-2,2'-bithiophene 97.0+%, TCI America™

Catalog No. O03835G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03835G 5 g
O03831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O03835G Supplier TCI America Supplier No. O03835G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 93164-73-9
Molecular Formula C16H22S2
Molecular Weight (g/mol) 278.472
InChI Key ZYEIXHZLTHTEOI-UHFFFAOYSA-N
PubChem CID 56973709
IUPAC Name 2-octyl-5-thiophen-2-ylthiophene
SMILES CCCCCCCCC1=CC=C(S1)C2=CC=CS2

Specifications

Melting Point 29°C
Color Yellow
Quantity 5 g
Formula Weight 278.47
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 5-n-Octyl-2,2′-bithiophene
TSCA No
Recommended Storage Freezer