accessibility menu, dialog, popup

UserName

NNH2NNH2

CAS RN 93127-75-4

IUPAC Name: [2,2'-bipyridine]-6,6'-diamine
Synonyms: [2,2'-bipyridine]-6,6'-diamine
Molecular Weight (g/mol): 186.22
Molecular Formula: C10H10N4
InChi Key: YKSWVQYWQSZDPR-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=N1)C1=NC(N)=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results