missing translation for 'onlineSavingsMsg'
Learn More

6,6'-Diamino-2,2'-bipyridyl 98.0+%, TCI America™

Catalog No. D2913250MG
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2913250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2913250MG Supplier TCI America Supplier No. D2913250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory reagent with 98.0% minimum purity.

Chemical Identifiers

CAS 93127-75-4
Molecular Formula C10H10N4
Molecular Weight (g/mol) 186.218
MDL Number MFCD04117918
InChI Key YKSWVQYWQSZDPR-UHFFFAOYSA-N
Synonym 2,2′C-Bipyridine-6,6′C-diamine
PubChem CID 10561426
IUPAC Name 6-(6-aminopyridin-2-yl)pyridin-2-amine
SMILES C1=CC(=NC(=C1)N)C2=NC(=CC=C2)N

Specifications

Melting Point 186°C
Quantity 250 mg
Formula Weight 186.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6,6′-Diamino-2,2′-bipyridyl