accessibility menu, dialog, popup

UserName

CAS RN 93-61-8

IUPAC Name: N-methyl-N-phenylformamide
Synonyms: N-methylformanilide
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: JIKUXBYRTXDNIY-UHFFFAOYSA-N
SMILES: CN(C=O)C1=CC=CC=C1
Boiling Point 
  • (4)
  • (3)
  • (4)
Molecular Weight (g/mol) 
  • (7)
  • (4)
Percent Purity 
  • (3)
  • (8)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (3)
  • (8)
  • (4)
  • (3)
  • (4)
  • (7)
  • (4)
  • (3)
  • (8)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (7)