CAS RN 93-26-5
CAS RN 93-26-5
IUPAC Name:
N-(2-methoxyphenyl)acetamide
Synonyms:
O-methoxyacetanilide
Molecular Weight (g/mol):
165.19
Molecular Formula:
C9H11NO2
InChi Key:
FGOFNVXHDGQVBG-UHFFFAOYSA-N
SMILES:
COC1=CC=CC=C1NC(C)=O