accessibility menu, dialog, popup

UserName

CH3NHNOOCH3

CAS RN 92945-27-2

IUPAC Name: ethyl 5-propyl-1H-pyrazole-3-carboxylate
Synonyms: ethyl 5-propyl-1H-pyrazole-3-carboxylate
Molecular Weight (g/mol): 182.22
Molecular Formula: C9H14N2O2
InChi Key: SUALHSUMUQQLJP-UHFFFAOYSA-N
SMILES: CCCC1=CC(=NN1)C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results