accessibility menu, dialog, popup

UserName

CAS RN 929-77-1

IUPAC Name: methyl docosanoate
Synonyms: methyl behenate
Molecular Weight (g/mol): 354.62
Molecular Formula: C23H46O2
InChi Key: QSQLTHHMFHEFIY-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCC(=O)OC
Percent Purity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)