missing translation for 'onlineSavingsMsg'
Learn More

Methyl behenate, Analytical Standard, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015918
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015918 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015918 Supplier Merck Emd Millipore Supplier No. 119405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 929-77-1
Physical Form Neat
Linear Formula CH3(CH2)20 COOCH3
MDL Number MFCD00009347
Melting Point 54°C to 56°C
Synonym Behenic acid methyl ester; Docosanoic acid methyl ester; Methyl docosanoate
Quantity 5 g
Formula Weight 354.61
Percent Purity ≥99.0% (GC)
Grade Analytical Standard