missing translation for 'onlineSavingsMsg'
Learn More

Methyl Behenate 96.0+%, TCI America™

Catalog No. B174810G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B174810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B174810G Supplier TCI America Supplier No. B174810G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 929-77-1
Molecular Formula C23H46O2
Molecular Weight (g/mol) 354.62
MDL Number MFCD00009347
InChI Key QSQLTHHMFHEFIY-UHFFFAOYSA-N
Synonym methyl behenate, docosanoic acid, methyl ester, behenic acid methyl ester, docosanoic acid methyl ester, behenic acid, methyl ester, unii-04kbo9r771, behenic methyl ester, kemester 9022, docosanoic acid methyl, acmc-209rjf
PubChem CID 13584
IUPAC Name methyl docosanoate
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 55°C
Color White
Boiling Point 393°C
Quantity 10 g
Formula Weight 354.62
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Behenate