accessibility menu, dialog, popup

UserName

CAS RN 928-80-3

IUPAC Name: decan-3-one
Synonyms: 3-decanone
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: XJLDYKIEURAVBW-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results