missing translation for 'onlineSavingsMsg'
Learn More

3-Decanone 98.0+%, TCI America™

Catalog No. D116225ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D116225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D116225ML Supplier TCI America Supplier No. D116225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 928-80-3
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00009554
InChI Key XJLDYKIEURAVBW-UHFFFAOYSA-N
Synonym 3-decanone, ethyl heptyl ketone, unii-cbt4991prj, ethyl n-heptyl ketone, cbt4991prj, acmc-209ri7, 3-decanone, fg, unii-3z25263nt0 component
PubChem CID 13576
ChEBI CHEBI:87309
IUPAC Name decan-3-one
SMILES CCCCCCCC(=O)CC

Specifications

Melting Point -4°C
Color Yellow
Boiling Point 211°C
Quantity 25 mL
UN Number 1224
Formula Weight 156.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Decanone