CAS RN 927-49-1
CAS RN 927-49-1
IUPAC Name:
undecan-6-one
Synonyms:
6-undecanone
Molecular Weight (g/mol):
170.3
Molecular Formula:
C11H22O
InChi Key:
ZPQAKYPOZRXKFA-UHFFFAOYSA-N
SMILES:
CCCCCC(=O)CCCCC
1
–
1
of
1
results
1
6-Undecanone 98.0+%, TCI America™
CAS: 927-49-1 Molecular Formula: C11H22O Molecular Weight (g/mol): 170.296 MDL Number: MFCD00009516 InChI Key: ZPQAKYPOZRXKFA-UHFFFAOYSA-N Synonym: 6-undecanone,dipentyl ketone,amyl ketone,diamyl ketone,pentyl ketone,6-oxoundecane,6undecanone,di-n-amyl ketone,ketones, c11,c11 ketones PubChem CID: 13561 IUPAC Name: undecan-6-one SMILES: CCCCCC(=O)CCCCC