missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Undecanone 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 226°C |
| Quantity | 25 mL |
| Formula Weight | 170.30 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 6-Undecanone |
Safety and Handling
| EINECSNumber | (2)-2475 |
| RTECSNumber | YQ2828000 |
| TSCA | Yes |