accessibility menu, dialog, popup

UserName

H3COOO

CAS RN 924-44-7

IUPAC Name: ethyl 2-oxoacetate
Synonyms: ethyl glyoxylate
Molecular Weight (g/mol): 102.09
Molecular Formula: C4H6O3
InChi Key: DBPFRRFGLYGEJI-UHFFFAOYSA-N
SMILES: CCOC(=O)C=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)