missing translation for 'onlineSavingsMsg'
Learn More

Ethyl glyoxalate, technical, ∼50% in toluene

Catalog No. AC425765000
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC425765000 500 g
AC425760010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC425765000 Supplier Thermo Scientific Chemicals Supplier No. 425765000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 924-44-7 | C4H6O3 | 102.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 924-44-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
InChI Key DBPFRRFGLYGEJI-UHFFFAOYSA-N
Synonym ethyl glyoxalate, ethyl glyoxylate, ethyl oxoacetate, glyoxylic acid ethyl ester, acetic acid, oxo-, ethyl ester, ethyl glyoxalate solution, unii-ixx9mq1j1i, acetic acid, 2-oxo-, ethyl ester, ixx9mq1j1i, 2-oxoacetic acid, ethyl ester
PubChem CID 70211
ChEBI CHEBI:53275
IUPAC Name ethyl 2-oxoacetate
SMILES CCOC(=O)C=O

Specifications

Density 1.0320g/mL
Boiling Point 110°C
Flash Point 7°C
Packaging Glass Bottle
Linear Formula HCOCO2CH2CH3
Quantity 500 g
Specific Gravity 1.032
Formula Weight 102.09
Grade Technical
Chemical Name or Material Ethyl glyoxalate, Approx. 50% in toluene
Hazard Category GHS Signal Word: Danger
EINECSNumber 213-105-3

RUO – Research Use Only