accessibility menu, dialog, popup

UserName

CAS RN 924-16-3

IUPAC Name: dibutyl(nitroso)amine
Synonyms: N-nitrosodibutylamine
Molecular Weight (g/mol): 158.25
Molecular Formula: C8H18N2O
InChi Key: YGJHZCLPZAZIHH-UHFFFAOYSA-N
SMILES: CCCCN(CCCC)N=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results