CAS RN 920-39-8
IUPAC Name:
bromo(propan-2-yl)magnesium
Synonyms:
bromo(isopropyl)magnesium
Molecular Weight (g/mol):
147.3
Molecular Formula:
C3H7BrMg
InChi Key:
UCJIVFREPMUZDC-UHFFFAOYSA-M
SMILES:
CC(C)[Mg]Br
1
–
1
of
1
results
1
Promotions Available
Isopropylmagnesium bromide, 3M solution in 2-MeTHF, AcroSeal™
Isopropylmagnesium bromide, 3M solution in 2-MeTHF, AcroSeal™
CAS: 920-39-8 Molecular Formula: C3H7BrMg Molecular Weight (g/mol): 147.30 MDL Number: MFCD00672110 InChI Key: UCJIVFREPMUZDC-UHFFFAOYSA-M Synonym: isopropylmagnesium bromide,magnesium, bromo 1-methylethyl,isopropylmagnesiumbromide,bromo 1-methylethyl magnesium,iprmgbr,i-prmgbr,bromoisopropylmagnesium,i-propylmagnesiumbromide,i-propylmagnesium bromide,bromo-isopropyl-magnesium,grignard reagent PubChem CID: 101909 IUPAC Name: magnesium;propane;bromide SMILES: CC(C)[Mg]Br