missing translation for 'onlineSavingsMsg'
Learn More

Isopropylmagnesium bromide, 3M solution in 2-MeTHF, AcroSeal™

Catalog No. AC426781000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426781000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC426781000 Supplier Thermo Scientific Chemicals Supplier No. 426781000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 920-39-8 | C3H7BrMg | 147.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 920-39-8 , 96-47-9
Molecular Formula C3H7BrMg
Molecular Weight (g/mol) 147.30
MDL Number MFCD00672110
InChI Key UCJIVFREPMUZDC-UHFFFAOYSA-M
Synonym isopropylmagnesium bromide, magnesium, bromo 1-methylethyl, isopropylmagnesiumbromide, bromo 1-methylethyl magnesium, iprmgbr, i-prmgbr, bromoisopropylmagnesium, i-propylmagnesiumbromide, i-propylmagnesium bromide, bromo-isopropyl-magnesium, grignard reagent
PubChem CID 101909
IUPAC Name magnesium;propane;bromide
SMILES CC(C)[Mg]Br

Specifications

Color Brown or Gray
Density 1.0800g/mL
Flash Point −11°C
Packaging AcroSeal™ Glass Bottle
Linear Formula (CH3)2CHMgBr
Quantity 100 mL
UN Number 3399
Specific Gravity 1.08
Solubility Information Solubility in water: reacts
Formula Weight 147.31
Physical Form Solution
Chemical Name or Material Isopropylmagnesium bromide, 3M solution in 2-Methyltetrahydrofuran
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritati
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 213-056-8

RUO – Research Use Only