CAS RN 92-83-1
IUPAC Name:
9H-xanthene
Synonyms:
xanthan gum dibenzo[a,e]pyran
Molecular Weight (g/mol):
182.22
Molecular Formula:
C13H10O
InChi Key:
GJCOSYZMQJWQCA-UHFFFAOYSA-N
SMILES:
C1C2=CC=CC=C2OC2=CC=CC=C12
1
–
1
of
1
results
1
Xanthene 98.0+%, TCI America™
CAS: 92-83-1 Molecular Formula: C13H10O Molecular Weight (g/mol): 182.222 MDL Number: MFCD00005055 InChI Key: GJCOSYZMQJWQCA-UHFFFAOYSA-N Synonym: xanthene,xanthan,10h-9-oxaanthracene,dibenzo a,e pyran,dibenzopyran, tricyclic,9h-xanthen,xanthane,9h-xanthen-9-yl,acmc-209rhu PubChem CID: 7107 ChEBI: CHEBI:10057 IUPAC Name: 9H-xanthene SMILES: C1C2=CC=CC=C2OC3=CC=CC=C31