accessibility menu, dialog, popup

UserName

ClSNH

CAS RN 92-39-7

IUPAC Name: 2-chloro-10H-phenothiazine
Synonyms: 10H-phenothiazine, 2-chloro-
Molecular Weight (g/mol): 233.71
Molecular Formula: C12H8ClNS
InChi Key: KFZGLJSYQXZIGP-UHFFFAOYSA-N
SMILES: ClC1=CC=C2SC3=CC=CC=C3NC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results