accessibility menu, dialog, popup

UserName

CAS RN 918-00-3

H3COClClCl

CAS RN 918-00-3

IUPAC Name: 1,1,1-trichloropropan-2-one
Synonyms: 1,1,1-trichloroacetone
Molecular Weight (g/mol): 161.41
Molecular Formula: C3H3Cl3O
InChi Key: SMZHKGXSEAGRTI-UHFFFAOYSA-N
SMILES: CC(=O)C(Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results