missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trichloroacetone 95.0+%, TCI America™

Catalog No. T137625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T137625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T137625G Supplier TCI America Supplier No. T137625G
Only null left

Chemical Identifiers

CAS 918-00-3
Molecular Formula C3H3Cl3O
Molecular Weight (g/mol) 161.41
MDL Number MFCD00018829
InChI Key SMZHKGXSEAGRTI-UHFFFAOYSA-N
Synonym 1,1,1-trichloroacetone, trichloroacetone, 1,1,1-trichloropropanone, 1,1,1-trichloro-2-propanone, 2-propanone, 1,1,1-trichloro, 2-propanone, trichloro, unii-nw27zg5lda, ccris 7196, alpha,alpha,alpha-trichloroacetone, nw27zg5lda
PubChem CID 13514
IUPAC Name 1,1,1-trichloropropan-2-one
SMILES CC(=O)C(Cl)(Cl)Cl

Specifications

Color Yellow
Boiling Point 134°C
Quantity 25 g
Formula Weight 161.41
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1-Trichloroacetone
RTECSNumber UC3839000
TSCA No