accessibility menu, dialog, popup

UserName

CAS RN 91419-52-2

IUPAC Name: tert-butyl 4-cyanopiperidine-1-carboxylate
Synonyms: tert-butyl 4-cyanopiperidine-1-carboxylate
Molecular Weight (g/mol): 210.28
Molecular Formula: C11H18N2O2
InChi Key: UQADQTBQNVARAP-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(CC1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results